missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CACNA2D1 Protein
A DNA sequence encoding the Homo sapiens (Human) CACNA2D1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5745-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000713.2 | |
CACNA2D1 Protein | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
781 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
CACNA2D1 | |
Recombinant Protein | |
Yeast | |
QPKPIGVGIP TINLRKRRPN IQNPKSQEPV TLDFLDAELE NDIKVEIRNK MIDGESGEKT FRTLVKSQDE RYIDKGNRTY TWTPVNGTDY SLALVLPTYS FYYIKAKLEE TITQARYSET LKPDNFEESG YTFIAPRDYC N |