missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C20ORF20 Protein
A cDNA sequence encoding the C20ORF20 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11237-10ug
Additional Details : Weight : 0.01000kg
Specifications
C20ORF20 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGEAEVGGGG AAGDKGPGEA ATSPAEETVV WSPEVEVCLF HAMLGHKPVG VNRHFHMICI RDKFSQNIGR QVPSKVIWDH LSTMYDMQAL HESEILPFPN PERNFVLPEE IIQEVREGKV MIEEEMKEEM KEDVDPHNGA DDVFSSSGSL GKASEKSSKD KEKNSSDLGC KEGADKRKRS RVTDKVLTAN SNPSSPSAAK RRRT | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C20ORF20 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol and 0.2M NaCl. |