missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1QTNF9 Protein

Product Code. 15906578
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15906578

Brand: enQuireBio™ QP112362ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the C1QTNF9 was constructed and used to recombinantly synthesize the protein.

Specifications

Name C1QTNF9 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MKHHHHHHAS QDTCRQGHPG IPGNPGHNGL PGRDGRDGAK GDKGDAGEPG RPGSPGKDGT SGEKGERGAD GKVEAKGIKG DQGSRGSPGK HGPKGLAGPM GEKGLRGETG PQGQKGNKGD VGPTGPEGPR GNIGPLGPTG LPGPMGPIGK PGPKGEAGPT GPQGEPGVRG IRGWKGDRGE KGKIGETLVL PKSAFTVGLT VLSKFPSSDM PIKFDKILYN EFNHYDTAAG KFTCHIAGVY YFTYHITVFS RNVQVSLVKN GVKILHTKDA YMSSEDQASG GIVLQLKLGD EVWLQVTGGE RFNGLFADED DDTTFTGFLL FSSP
Buffer C1QTNF9 Human was filtered (0.4microm) and lyophilized from 0.5 mg/ml supplied in 0.03M Acetate buffer, pH 4.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.