missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QTNF8 Protein
A cDNA sequence encoding the C1QTNF8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11235-2ug
Additional Details : Weight : 0.01000kg
Specifications
C1QTNF8 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS WPGLPRRPCV HCCRPAWPPG PYARVSDRDL WRGDLWRGLP RVRPTIDIEI LKGEKGEAGV RGRAGRSGKE GPPGARGLQG RRGQKGQVGP PGAACRRAYA AFSVGRRAYA AFSVGRREGL HSSDHFQAVP FDTELVNLDG AFDLAAGRFL CTVPGVYFLS LNVHTWNYKE TYLHIMLNRR PAAVLYAQPS ERSVMQAQSL MLLLAAGDAV WVRMFQRDRD NAIYGEHGDL YITFSGHLVK PAAEL |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C1QTNF8 filtered (0.4 microm) solution at a concentration of 0.5 mg/ml in 0.03M Acetate buffer pH 4.0. |