missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human C1qTNF5/CTRP5 His Protein

Product Code. 18607621 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µg
50µg
This item is not returnable. View return policy

Product Code. 18607621

Brand: Novus Biologicals™ NBP27750150ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.25 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH8.5), 30% glycerol, 0.2 M NaCl
Gene ID (Entrez) 114902
Molecular Weight (g/mol) MolecularWeight-theroretical: 26.4 kDa
Quantity 50 μg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Endotoxin Concentration Less Than 1.0 EU / 1 μg of protein (determined by LAL method)
Gene Alias C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3
Gene Symbol C1QTNF5
Research Category Vision
Purity or Quality Grade >85%, by SDS-PAGE
Protein C1qTNF5/CTRP5
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.