missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1q Protein

Product Code. 15946548
Click to view available options
Quantity:
1 mg
10 mg
200 μg
Unit Size:
10mg
1mg
200µg
This item is not returnable. View return policy

Product Code. 15946548

Brand: enQuireBio™ QP1122810mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the C1q was constructed and used to recombinantly synthesize the protein.

Specifications

Name C1q Protein
Quantity 10 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Buffer Lyophilized from 0.01M Na2 EDTA solution containing 0.3M NaCl pH 7.5.
Purity or Quality Grade Greater than 96.0% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.