missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C1q Protein

Product Code. 15966548
Change view
Click to view available options
Quantity:
1 mg
10 mg
200 μg
Unit Size:
10mg
1mg
200µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15966548 200 μg 200µg
15956548 1 mg 1mg
15946548 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15966548 Supplier enQuireBio™ Supplier No. QP11228200ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the C1q was constructed and used to recombinantly synthesize the protein.

Specifications

Name C1q Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human
Protein Tag Untagged
Buffer Lyophilized from 0.01M Na2 EDTA solution containing 0.3M NaCl pH 7.5.
Purity or Quality Grade Greater than 96.0% as determined by SDS-PAGE.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.