missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C11ORF31 Protein
A cDNA sequence encoding the C11ORF31 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11221-5ug
Additional Details : Weight : 0.01000kg
Specifications
C11ORF31 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPRGRK RKAEAAVVAV AEKREKLANG GEGMEEATVV IEHCTSCRVY GRNAAALSQA LRLEAPELPV KVNPTKPRRG SFEVTLLRPD GSSAELWTGI KKGPPRKLKF PEPQEVVEEL KKYLS | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The C11ORF31 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |