missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-X-C motif chemokine 17 Protein
Click to view available options
Quantity:
100 μg
25 μg
250 μg
5 μg
Unit Size:
100µg
250µg
25µg
5µg
Specifications
Specifications
| Accession Number | NP_940879.1 |
| Gene ID (Entrez) | 284340 |
| Name | C-X-C motif chemokine 17 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than 1.0 EU/ug as determined by LAL method. |
| Gene Symbol | CXCL17 |
| Protein Length | Partial (see sequence information for more details). |
| Biological Activity | The ED50 as determined by its ability to induce VEGF expression using murine endothelial cells is less than 5.0 ug/ml, corresponding to a specific activity of > 200 IU/mg. |
| Product Type | Recombinant Protein |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction