missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-X-C motif chemokine 17 Protein

Product Code. 15903128
Click to view available options
Quantity:
100 μg
25 μg
250 μg
5 μg
Unit Size:
100µg
250µg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15903128

Brand: enQuireBio™ QP10220ec25ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human C-X-C motif chemokine 17, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_940879.1
Gene ID (Entrez) 284340
Name C-X-C motif chemokine 17 Protein
Quantity 25 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol CXCL17
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by its ability to induce VEGF expression using murine endothelial cells is less than 5.0 ug/ml, corresponding to a specific activity of > 200 IU/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence SSLNPGVARG HRDRGQASRR WLQEGGQECE CKDWFLRAPR RKFMTVSGLP KKQCPCDHFK GNVKKTRHQR HHRKPNKHSR ACQQFLKQCQ LRSFALPL
Buffer 0.2 um filtered PBS, pH 7.4, lyophilized
Purity or Quality Grade Greater than 96% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.