missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human C-Reactive Protein/CRP Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 .
Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 with BSA as a carrier protein.
Unit Size:
200µg
Description
The Recombinant Human C-Reactive Protein/CRP Protein is derived from NS0. The Recombinant Human C-Reactive Protein/CRP Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in Tris-HCl, NaCl and CaCl 2 . |
| Gene ID (Entrez) | 1401 |
| Molecular Weight (g/mol) | 23 kDa (monomer) |
| Name | C-Reactive Protein/CRP |
| Quantity | 200 μg |
| Source | Mouse myeloma cell line,NS0-derived human C-Reactive Protein/CRP protein Phe17-Pro224 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Homopentamer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction