missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Recombinant Human BTN2A1 Fc Chimera Protein, CF

Product Code. 30023103
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30023103 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30023103 Supplier R&DSystems™ Supplier No. 11521BT050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Recombinant Human BTN2A1 Fc Chimera binds to biotinylated DC-SIGN Fc Chimera with a ED50 of 1.00-10.0 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7KYR7.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 11120
Molecular Weight (g/mol) M.W.-observed: 60-80 kDa, under reducing conditions., M.W.-theroretical: 51 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human BTN2A1 protein Human BTN2A1 (Gln29-Ala248) (N-terminus) Accession # Q7KYR7.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias BK14H9.1, BT2.1, BTF1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein BTN2A1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.