missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BTC / Betacellulin Protein
A DNA sequence encoding the Homo sapiens (Human) BTC / Betacellulin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5728-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001303892.1 | |
BTC / Betacellulin Protein | |
Research Use Only | |
BTC | |
Recombinant Protein | |
Yeast | |
DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY LRGDRGQILV ICLIAVMVVF IILVIGVCTC CHPLRKRRKR KKKEEEMETL GKDITPINED IEETNIA | |
Greater than 90% as determined by SDS-PAGE. |
685 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |