missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BTC / Betacellulin Protein

A DNA sequence encoding the Homo sapiens (Human) BTC / Betacellulin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5728-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967309

  • £393.00 / 100µg
Estimated Shipment: 05-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001303892.1
BTC / Betacellulin Protein
Research Use Only
BTC
Recombinant Protein
Yeast
DGNSTRSPET NGLLCGDPEE NCAATTTQSK RKGHFSRCPK QYKHYCIKGR CRFVVAEQTP SCVCDEGYIG ARCERVDLFY LRGDRGQILV ICLIAVMVVF IILVIGVCTC CHPLRKRRKR KKKEEEMETL GKDITPINED IEETNIA
Greater than 90% as determined by SDS-PAGE.
685
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers