missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bruton Tyrosine Kinase / BTK Kinase Protein

Product Code. 16077451
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16077451

Brand: enQuireBio™ QP9013ye1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Bruton Tyrosine Kinase / BTK Kinase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000052.1
Gene ID (Entrez) 695
Name Bruton Tyrosine Kinase / BTK Kinase Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Btk
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag His
Sequence AAVILESIFL KRSQQKKKTS PLNFKKRLFL LTVHKLSYYE YDFERGRRGS KKGSIDVEKI TCVETVVPEK NPPPERQIPR RGEESSEMEQ ISIIERFPYP FQVVYDEGPL YVFSPTEELR KRWIHQLKNV IRYNSDLVQK YHPCFWIDGQ YLCCSQTAKN AMGCQILENR NGSLKPGSSH RKTKKPLPPT PEEDQILKKP LPPEPAAAPV STSELKKVVA LYDYMPMNAN DLQLRKGDEY FILEESNLPW WRARDKNGQE GYIPSNYVTE AEDSIEMYEW YSKHMTRSQA EQLLKQEGKE GGFIVRDSSK AGKYTVSVFA KSTGDPQGVI RHYVVCSTPQ SQYYLAEKHL FSTIPELINY HQHNSAGLIS RLKYPVSQQN KNAPSTAGLG YGSWEIDPKD LTFLKELGTG QFGVVKYGKW RGQYDVAIKM IKEGSMSEDE FIEEAKVMMN LSHEKLVQLY GVCTKQRPIF IITEYMANGC LLNYLREMRH RFQTQQLLEM CKDVCEAMEY LESKQFLHRD LAARNCLVND QGVVKVSDFG LSRYVLDDEY TSSVGSKFPV RWSPPEVLMY SKFSSKSDIW AFGVLMWEIY SLGKMPYERF TNSETAEHIA QGLRLYRPHL ASEKVYTIMY SCWHEKADER PTFKILLSNI LDVMDEES
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.