missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRD3 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | BRD3 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSTATTV APAGIPATPG PVNPPPPEVS NPSKPGRKTN QLQYMQNVVV KTLWKHQFAW PFYQPVDAIK LNLPDYHKII KNPMDMGTIK KRLENNYYWS ASECMQDFNT MFTNCYIYNK PTDDIVLMAQ ALEKIFLQKV AQMPQEEVEL LPPAPKGKGR KPAAGAQSAG TQQVAAVSSV SPATPFQSVP PTVSQTPVIA ATPVPTITAN VTSVPVPPAA APPPPATPIV PVVPPTPPVV KKKGVKRKAD TTTPTTSAIT ASRSESPPPL SDPKQAKVVA RRESGGRPIK PPKKDLEDGE VPQHAGKKGK LSEHLRYCDS ILREMLSKKH AAYAWPFYKP VDAEALELHD YHDIIKHPMD LSTVKRKMDG REYPDAQGFA ADVRLMFSNC YKYNPPDHEV VAMARKLQDV FEMRFAKMP |
| Buffer | BRD3 protein solution (0.5 mg/ml) containing PBS buffer (pH 7.4) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction