missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BPI Protein

Product Code. 15906458
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15906458

Brand: enQuireBio™ QP111982ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the BPI was constructed and used to recombinantly synthesize the protein.

Specifications

Name BPI Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MRENMARGPC NAPRWASLMV LVAIGTAVTA AVNPGVVVRI SQKGLDYASQ QGTAALQKEL KRIKIPDYSD SFKIKHLGKG HYSFYSMDIR EFQLPSSQIS MVPNVGLKFS ISNANIKISG KWKAQKRFLK MSGNFDLSIE GMSISADLKL GSNPTSGKPT ITCSSCSSHI NSVHVHISKS KVGWLIQLFH KKIESALRNK MNSQVCEKVT NSVSSELQPY FQTLPVMTKI DSVAGINYGL VAPPATTAET LDVQMKGEFY SENHHNPPPF APPVMEFPAA HDRMVYLGLS DYFFNTAGLV YQEAGVLKMT LRDDMIPKES KFRLTTKFFG TFLPEVAKKF PNMKIQIHVS ASTPPHLSVQ PTGLTFYPAV DVQAFAVLPN SSLASLFLIG MHTTGSMEVS AESNRLVGEL KLDRLLLELK HSNIGPFPVE LLQDIMNYIV PILVLPRVNE KLQKGFPLPT PARVQLYNVV LQPHQNFLLF GADVVYK
Buffer BPI (1.15 mg/ml) is supplied in 25mM Sodium Acetate pH 4.6.
Purity or Quality Grade Greater than 70% as determined by SDS-PAGE and capillary electrophoresis.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.