missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BMP10 Protein

Product Code. 15974168
Change view
Click to view available options
Quantity:
1 μg
10 μg
5 μg
Unit Size:
10µg
1µg
5µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15974168 1 μg 1µg
15984168 5 μg 5µg
15964168 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15974168 Supplier enQuireBio™ Supplier No. QP105361ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the BMP10 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 27302
Name BMP10 Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol BMP10
Biological Activity The specific activity was determined by its ability to induce alkaline phosphatase production in the ATDC-5 cell line and is typically 4.0-6.0 ng/ml ED50.
Product Type Recombinant Protein
Cross Reactivity Human
Species Human (HEK293)
Protein Tag Untagged
Sequence NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALVHLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR
Buffer The BMP10 protein was lyophilized from 10mM Sodium Citrate, pH 2.7 and 75mM NaCl .
Purity or Quality Grade Greater than 95.0% as determined by analysis by SDS-PAGE and HPLC analysis.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.