missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BMP-8b Protein

Product Code. 18795847
Click to view available options
:
Quantity: 25μg
This item is not returnable. View return policy

Product Code. 18795847

Brand: R&D Systems™ 9316BP025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human BMP-8b Protein is derived from CHO. The Recombinant Human BMP-8b Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P34820
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in HCl
Gene ID (Entrez) 656
Molecular Weight (g/mol) 16 kDa
Name BMP-8b Recombinant Protein
Quantity 25μg
Source Chinese Hamster Ovary cell line,CHO-derived human BMP-8b protein Ala264-His402
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Common Name BMP-8; BMP-8b; bone morphogenetic protein 8b; MGC131757; OP-3
Biological Activity 0.7 to 3.5μg/mL
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.