missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-8a Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£361.76
Specifications
Gene ID (Entrez) | 656 |
---|---|
Molecular Weight (g/mol) | 15.8 kDa (monomer) |
Name | BMP-8a |
Quantity | 10μg |
Source | E. coli-derived human BMP-8a protein Ala264-His402 & Val265-His402 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18128390
|
R&D Systems™
1073-BP-010 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
£361.76
10µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18186950
|
R&D Systems™
1073-BP-010/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
£361.76
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human BMP-8a Protein is derived from E. coli. The Recombinant Human BMP-8a Protein has been validated for the following applications: Binding Activity.Specifications
656 | |
BMP-8a | |
E. coli-derived human BMP-8a protein Ala264-His402 & Val265-His402 | |
Disulfide-linked homodimer | |
Recombinant |
15.8 kDa (monomer) | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |