missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human BMP-7 Protein

Product Code. p-200055396 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18493662

Brand: Novus Biologicals™ NBP234933500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18075, P18075, P18075
Concentration LYOPH
For Use With (Application) Western Blot, ELISA, PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 30 % acetonitrile, 0.1 % TFA.
Gene ID (Entrez) 655
Molecular Weight (g/mol) 15.7 kDa
Quantity 500 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of BMP-7 as determined by LAL method.
Gene Alias BMP-7, bone morphogenetic protein 7, Eptotermin alfa, OP-1OP1, Osteogenic protein 1
Gene Symbol BMP7
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in 10 mM HAc to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Inhibitor Properties 1. Molecular standard (Western, ELISA) in studying secreted BMP-7; 2. Preparing antibodies for BMP-7 monomer; 3. Molecule standard in detecting secreted BMP-7 in reduced SDS-PAGE.
Research Category Biologically Active Proteins, Cytokine Research
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein BMP-7
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.