missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Accession Number | P12644, P12644, P12644 |
| Concentration | LYOPH |
| For Use With (Application) | Western Blot, ELISA, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 50 mM Na2CO3, 5 mM DTT, 0.02% Tween 20, pH 11.0. |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | 13.3 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of BMP-4 as determined by LAL method. |
| Gene Alias | BMP-2B, BMP2B1, BMP2BMCOPS6, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, OFC11, ZYME |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?