missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-4 (E. coli-expressed) Protein
Click to view available options
Quantity:
10 μg
50 μg
Unit Size:
10µg
50µg
Specifications
Specifications
| Accession Number | P12644 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 652 |
| Molecular Weight (g/mol) | 12 kDa |
| Name | BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME |
| Quantity | 10 μg |
| Source | E. coli-derived human BMP-4 protein Lys303-Arg408 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -70 °C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction