missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BMP2 protein is fully biologically active when compared to standard. The ED50 as determined by inducing alkaline phosphatase production of murine ATDC5 cells is less than 200 ng/mL, corresponding to a specific activity of >5.0 x 10^3 IU/mg.
Specifications
Specifications
| Accession Number | P12643, P12643, P12643 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution containing 10 mM sodium citrate pH 3.5. |
| Gene ID (Entrez) | 650 |
| Molecular Weight (g/mol) | 26.0 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of BMP-2 as determined by LAL method. |
| Gene Alias | BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?