missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-15 Protein
Description
The Recombinant Human BMP-15 Protein is derived from CHO. The Recombinant Human BMP-15 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein. |
| Gene ID (Entrez) | 9210 |
| Molecular Weight (g/mol) | 13.9 kDa |
| Name | BMP-15/GDF-9B |
| Quantity | 5μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human BMP-15/GDF-9B protein Gln268-Arg392 (Thr266Arg) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | BMP15, BMP-15, bone morphogenetic protein 15, GDF-9 B, Growth/differentiation factor 9 B, ODG2, POF4 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?