missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BIRC7 Protein

A cDNA sequence encoding the BIRC7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11164-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936348

  • £155.00 / 20µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BIRC7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGPKDSAKCL HRGPQPSHWA AGDGPTQERC GPRSLGSPVL GLDTCRAWDH VDGQILGQLR PLTEEEEEEG AGATLSRGPA FPGMGSEELRLASFYDWPLT AEVPPELLAA AGFFHTGHQD KVRCFFCYGG LQSWKRGDDP WTEHAKWFPS CQFLLRSKGR DFVHSVQETH SQLLGSWDPW EEPEDAAPVA PSVPASGYPE LPTPRREVQS ESAQEPGGVS PAEAQRAWWV LEPPGARDVE AQLRRLQEER TCKVCLDRAV SIVFVPCGHL VCAECAPGLQ LCPICRAPVR SRVRTFLS
Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The BIRC7 solution contains 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl, 2mM EDTA and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers