missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Bim His Protein

Product Code. 18071364 Shop All Bio Techne Products
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18071364

Brand: Novus Biologicals™ NBP19887350UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-138 of Human Bim The Recombinant Human Bim Protein is derived from E. coli. The Recombinant Human Bim Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43521
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer, pH8.0, 2M Urea, 20% glycerol 5mM DTT, 300mM NaCl
Gene ID (Entrez) 10018
Name Bim Protein
Quantity 50 μg
Source E. Coli
Immunogen BCL2L11, 1-138 aa Human, His tag, E.coli (NP_006529).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >80%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.