missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Biliverdin Reductase B/BLVRB Protein
Description
The Recombinant Human Biliverdin Reductase B/BLVRB Protein is derived from E. coli. The Recombinant Human Biliverdin Reductase B/BLVRB Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, Brij and Glycerol. |
| Gene ID (Entrez) | 645 |
| Molecular Weight (g/mol) | 23 kDa |
| Name | Biliverdin Reductase B/BLVRB |
| Quantity | 20 μg |
| Source | E. coli-derived human Biliverdin Reductase B/BLVRB protein Ala2-Gln206,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?