missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BID Protein
A DNA sequence encoding the Homo sapiens (Human) BID Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5718-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001187.1 | |
BID Protein | |
Research Use Only | |
BID | |
Recombinant Protein | |
E. coli | |
MDCEVNNGSS LRDECITNLL VFGFLQSCSD NSFRRELDAL GHELPVLAPQ WEGYDELQTD GNRSSHSRLG RIEADSESQE DIIRNIARHL AQVGDSMDRS IPPGLVNGLA LQLRNTSRSE EDRNRDLATA LEQLLQAYPR DMEKEKTMLV LALLLAKKVA SHTPSLLRDV FHTTVNFINQ NLRTYVRSLA RNGMD | |
Greater than 90% as determined by SDS-PAGE. |
637 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |