missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Betacellulin Protein

Product Code. 18777736
missing translation for 'orderingAttributeHoverText'
Quantity:
250 μg
missing translation for 'unitSize'
250µg
This item is not returnable. View return policy

Product Code. 18777736

missing translation for 'mfr': R&D Systems™ 261CE250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human Betacellulin Protein is derived from E. coli. The Recombinant Human Betacellulin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35070
For Use With (Application) Bioactivity
Gene ID (Entrez) 685
Molecular Weight (g/mol) 9 kDa
Quantity 250 μg
Source E. coli-derived human Betacellulin/BTC protein Asp32-Tyr111
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Symbol BTC
Specific Reactivity Betacellulin/BTC
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS containing at least 0.1% human or bovine serum albumin.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.