missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human beta-NGF Protein

Product Code. 18106280
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetic Acid.
Molecular Weight (g/mol):
13.5 kDa
13.5 kDa (monomer)
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18106280

Brand: R&D Systems™ 256GF100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human beta-NGF Protein is derived from NS0. The Recombinant Human beta-NGF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetic Acid with BSA as a carrier protein.
Gene ID (Entrez) 4803
Molecular Weight (g/mol) 13.5 kDa (monomer)
Name beta-NGF
Quantity 100 μg
Source Mouse myeloma cell line,NS0-derived human beta-NGF protein Ser122-Ala241
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.