missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human beta-Defensin 4/2 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
Description
Defensin beta 4 protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 0.1-100.0 ng/mL.
Specifications
Specifications
| Accession Number | Q8WTQ1 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 130 mM NaCl. |
| Gene ID (Entrez) | 1673 |
| Molecular Weight (g/mol) | 6.0 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of beta-Defensin 4/2 as determined by LAL method. |
| Gene Alias | Beta-defensin 2, DEFB-2, DEFB2DEFB102, DEFB4beta-defensin 4A, Defensin, beta 2Skin-antimicrobial peptide 1, defensin, beta 4, defensin, beta 4A, HBD-2, SAP1BD-2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction