missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human beta-Defensin 3 Protein

Product Code. 18324118
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324118 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324118 Supplier R&D Systems™ Supplier No. 4435BD050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its anti-microbial activity against E. coli. Ganz, T. et al. (2003) Nat. Rev. Immunol. 3:710. The ED50 for this effect is 7.5-30 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P81534
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl.
Gene ID (Entrez) 55894
Molecular Weight (g/mol) M.W. (theoretical): 5.2 kDa
Name beta-Defensin 3
Quantity 50 μg
Source E. coli-derived human beta-Defensin 3 protein Gly23-Lys67
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol DEFB103B
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein beta-Defensin 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.