missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-catenin-interacting protein 1

A DNA sequence encoding the Homo sapiens (Human) Beta-catenin-interacting protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8120-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16042771

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001012329.1
Beta-catenin-interacting protein 1
Research Use Only
CTNNBIP1
Recombinant Protein
E. coli
MNREGAPGKS PEEMYIQQKV RVLLMLRKMG SNLTASEEEF LRTYAGVVNS QLSQLPPHSI DQGAEDVVMA FSRSETEDRR Q
Greater than 80% as determined by SDS-PAGE.
56998
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers