missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL2 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | BCL2 Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | AHAGRTGYDN REIVMKYIHY KLSQRGYEWD AGDVGAAPPG AAPAPGIFSS QPGHTPHPAA SRDPVARTSP LQTPAAPGAA AGPALSPVPP VVHLTLRQAG DDFSRRYRRD FAEMSSQLHL TPFTARGRFA TVVEELFRDG VNWGRIVAFF EFGGVMCVES VNREMSPLVD NIALWMTEYL NRHLHTWIQD NGGWDAFVEL YGPSMRPLFD |
| Buffer | BCL2 is a 0.2microm filtered concentrated solution which contains 25 mM Tris-HCl, pH 8.0, 100mM NaCl, 1 mM DTT, 30 % Glycerol, and 0.01%Tween-80. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction