missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl XL Protein

A cDNA sequence encoding the Bcl XL was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11141-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956278

  • £155.00 / 50µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Bcl XL Protein
Research Use Only
Recombinant Protein
E. coli
The protein contains 10mM Tris-HCl pH 8.0, 1mM EDTA and 250mM NaCl.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers