missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Bcl-w Western Blot Standard Protein
Description
The Recombinant Human Bcl-w Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications
Specifications
| Gene ID (Entrez) | 599 |
| Name | Bcl-w Western Blot Standard |
| Quantity | 100 μL |
| Storage Requirements | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?