missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human BCL-W/BCL2L2 Protein

Product Code. 18402632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1000 μg
5 μg
500 μg
Unit Size:
1000µg
100µg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402632 500 μg 500µg
18461402 100 μg 100µg
18478581 1000 μg 1000µg
18434452 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18402632 Supplier Novus Biologicals™ Supplier No. NBP234895500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Test in Process.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92843, Q92843, Q92843
For Use With (Application) PAGE
Formulation 0.2 um filtered concentrated solution in 25 mM Hepes, pH 7.4, 100 mM KCl, 10 % Glycerol, 0.02 % Tween-80.
Gene ID (Entrez) 599
Molecular Weight (g/mol) 18.6 kDa
Quantity 500 μg
Storage Requirements Store at -20C. Avoid freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of BCL-W/BCL2L2 as determined by LAL method.
Gene Alias Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2
Gene Symbol BCL2L2
Research Category Apoptosis, Cancer
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein BCL-W/BCL2L2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.