missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Test in Process.
Specifications
Specifications
| Accession Number | Q92843, Q92843, Q92843 |
| For Use With (Application) | PAGE |
| Formulation | 0.2 um filtered concentrated solution in 25 mM Hepes, pH 7.4, 100 mM KCl, 10 % Glycerol, 0.02 % Tween-80. |
| Gene ID (Entrez) | 599 |
| Molecular Weight (g/mol) | 18.6 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20C. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of BCL-W/BCL2L2 as determined by LAL method. |
| Gene Alias | Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2-like 2, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 |
| Gene Symbol | BCL2L2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?