missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bcl 2 (minus BH1 domain) Protein
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
Specifications
Specifications
| Name | Bcl 2 (minus BH1 domain) Protein |
| Quantity | 500 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Untested |
| Product Type | Recombinant Protein |
| Cross Reactivity | Usutu virus |
| Species | Human |
| Protein Tag | His |
| Buffer | DPBS pH 7.4. |
| Purity or Quality Grade | >95% as determined by SDS-PAGE. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction