missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl 2 (minus BH1 domain) Protein

A cDNA sequence encoding the Bcl 2 (minus BH1 domain) was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10491-HIS-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954028

  • £99.15 / 2µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Bcl 2 (minus BH1 domain) Protein
Research Use Only
Recombinant Protein
E. coli
MAHAGRTGYD NREIVMKYIH YKLSQRGYEW DAGDVGAAPP GAAPAPGIFS SQPGHTPHPA ASRDPVARTS PLQTPAAPGA AAGPALSPVP PVVHLTLRQA GDDFSRRYRR DFAEMSSQLH LTPFTARGRF ATVVEELFRD GVNWGRIVAF FEFGGVMCVE SVNREMSPLV DNIALWMTEY LNRHLHTWIQ DNGGWDAFVE LYGPSMRPLF DFSWLSLKTL LSLALVGACI TLGAYLGHK
Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The protein contains 10mM Tris-HCL pH 8.0, 1mM EDTA and 250mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers