missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BASP1 Protein

A cDNA sequence encoding the BASP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11127-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936238

  • £99.15 / 2µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

10409
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMGGKLSK KKKGYNVNDE KAKEKDKKAE GAATEEEGTP KESEPQAAAE PAEAKEGKEK PDQDAEGKAE EKEGEKDAAA AKEEAPKAEP EKTEGAAEAK AEPPKAPEQE QAAPGPAAGG EAPKAAEAAA APAESAAPAA GEEPSKEEGE PKKTEAPAAP AAQETKSDGA PASDSKPGSS EAAPSSKETP AATEAPSSTP KAQGPAASAE EPKPVEAPAA NSDQTVTVKE
Greater than 95.0% as determined by SDS-PAGE.
BASP1 Protein
Research Use Only
BASP1
Human
His
The BASP1 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers