missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAG1 Protein
A cDNA sequence encoding the BAG1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11124-5ug
Additional Details : Weight : 0.01000kg
Specifications
BAG1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MNRSQEVTRD EESTRSEEVT REEMAAAGLT VTVTHSNEKH DLHVTSQQGS SEPVVQDLAQ VVEEVIGVPQ SFQKLIFKGK SLKEMETPLS ALGIQDGCRV MLIGKKNSPQ EEVELKKLKH LEKSVEKIAD QLEELNKELT GIQQGFLPKD LQAEALCKLD RRVKATIEQF MKILEEIDTL ILPENFKDSR LKRKGLVKKV QAFLAECDTV EQNICQETER LQSTNFALAE | |
Greater than 90% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
BAG1 Human solution containing 20mM Tris-HCl pH 7.5, 0.1M NaCl and 10% glycerol. |