missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BAG1 Protein

A cDNA sequence encoding the BAG1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11124-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15926228

  • £2490.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BAG1 Protein
Research Use Only
Recombinant Protein
E. coli
MNRSQEVTRD EESTRSEEVT REEMAAAGLT VTVTHSNEKH DLHVTSQQGS SEPVVQDLAQ VVEEVIGVPQ SFQKLIFKGK SLKEMETPLS ALGIQDGCRV MLIGKKNSPQ EEVELKKLKH LEKSVEKIAD QLEELNKELT GIQQGFLPKD LQAEALCKLD RRVKATIEQF MKILEEIDTL ILPENFKDSR LKRKGLVKKV QAFLAECDTV EQNICQETER LQSTNFALAE
Greater than 90% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
BAG1 Human solution containing 20mM Tris-HCl pH 7.5, 0.1M NaCl and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers