missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BAFF Receptor protein is fully biologically active when compared to standard. The ED50 as determined by its ability to block BAFF induced mouse splenocyte survival is 1.0-5.0 μg/mL in the presence of 1.0 μg/mL of rHuBAFF.
Specifications
Specifications
| Accession Number | Q96RJ3, Q96RJ3, Q96RJ3 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 8.0, 500 mM NaCl. |
| Gene ID (Entrez) | 115650 |
| Molecular Weight (g/mol) | 7.8 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of BAFFR/TNFRSF13C as determined by LAL method. |
| Gene Alias | BAFF Receptor/TNFRSF13C, BAFFR, BAFF-R, BROMIX, CD268, CVID4, prolixin, Tumor necrosis factor receptor superfamily member 13C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?