missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BAFF/BLyS/TNFSF13B Protein

Product Code. 18730169
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18730169

Brand: R&D Systems™ 2149BF01M/CF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BAFF/BLyS/TNFSF13B Protein is derived from NS0. The Recombinant Human BAFF/BLyS/TNFSF13B Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y275
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Gene ID (Entrez) 10673
Molecular Weight (g/mol) 19.6kDa
Quantity 1 mg
Source Mouse myeloma cell line,NS0-derived human BAFF/BLyS/TNFSF13B protein, (N-terminus) MHHHHHHHHHH GGGSGGGSGGGS IEGR Human BAFF (Ala134-Leu285) Accession &Num; Q9Y275 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Symbol TNFSF13B
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 100μg/mL in sterile PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.