missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BAALC Protein
A cDNA sequence encoding the BAALC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11121-1mg
Additional Details : Weight : 0.01000kg
Specifications
BAALC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGCGGSRADA IEPRYYESWT RETESTWLTY TDSDAPPSAA APDSGPEAGG LHSGMLEDGL PSNGVPRSTA PGGIPNPEKK TNCETQCPNP QSLSSGPLTQ KQNGLQTTEA KRDAKRMPAK EVTINVTDSI QQMDRSRRIT KNCVN | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein solution contains 20mM Tris buffer pH 8.0, 1mM DTT, 20% glycerol and 2mM EDTA. |