missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-H6 Fc Chimera Biotinylated Protein

Product Code. 18389447
Change view
Click to view available options
Quantity:
50 μg
Packungsgröße:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18389447 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18389447 Lieferant R&D Systems™ Lieferanten-Nr. BT7144050

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human NKp30 Fc Chimera (Catalog # 1849-NK ) is immobilized at 1 μg/mL, 100 μL/well, the concentration of Biotinylated Recombinant Human B7-H6 Fc Chimera that produces 50% of the optimal binding response is 15-75 ng/mL. Measured by its ability to induce IFN-gamma secretion by NK-92 human natural killer lymphoma cells. Brandt, C.S. et al. (2009) J. Exp. Med. 206:1495. The ED50 for this effect is 0.6-3 μg/mL
TRUSTED_SUSTAINABILITY

Spezifikation

Accession Number NP_001189368
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 374383
Molecular Weight (g/mol) M.W. (Observed): 74-85 kDa, reducing conditions; M.W. (theoretical): 53 kDa (unlabeled)
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived human B7-H6 protein Human B7-H6 (Met1-Ser262) (N-terminus) Accession # NP_001189368 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Biotin
Cross Reactivity B7-H6
Species Human
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H6
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.