missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-H3 Fc Chimera Protein

Product Code. 18349617
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349617 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349617 Supplier R&D Systems™ Supplier No. 1027B3100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Measured by its ability to inhibit anti-CD3-induced proliferation of stimulated human T cells. The ED50 for this effect is 3-12 μg/mL. Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_079516
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 80381
Molecular Weight (g/mol) M.W. (Observed): 70-80 kDa, reducing conditions; M.W. (theoretical): 50 kDa (monomer)
Name B7-H3 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived human B7-H3 protein Human B7-H3 (Leu29-Pro245) (N-terminus) Accession # NP_079516 DIEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol CD276
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H3/CD276
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.