missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-1/CD80 Fc Alexa Fluor™ 647 Protein

Product Code. 18768256
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768256 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18768256 Supplier R&D Systems™ Supplier No. AFR10133020
  • ONLINE EXCLUSIVE PRICE
    £474.00
    £421.05 / 20µg
    Save £52.95
    11% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fc Chimera

Measured by flow cytometry for its ability to bind anti-human B7-1 Monoclonal Antibody conjugated beads. The concentration of Recombinant Human B7-1 Fc-tag Alexa Fluor™ 647 (Catalog # AFR10133) that produces 50% of the binding response is 2.00-20.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P33681.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 941
Molecular Weight (g/mol) M.W.-observed: 66-80 kDa, under reducing conditions., M.W.-theroretical: 50.5 kDa
Quantity 20 μg
Source Chinese Hamster Ovary cell line, CHO-derived human B7-1/CD80 protein Human B7-1/CD80 (Val35-Asn242) (C-terminus) Accession # P33681.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus)
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80
Conjugate Alexa Fluor 647
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-1/CD80
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.